Ethyl-3-piperidinecarboxylate ≥99.9%

1233005-2 1233005-3 1233005-1
103535-318EA 38.46 USD
103535-318 103535-320 103536-280
Ethyl-3-piperidinecarboxylate ≥99.9%
Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MDL Number: MFCD00005991
CAS Number: 5006-62-2

Order Now

Specification Test Results

Purety 99.9%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR