4-Bromo-2-fluoroanisole ≥98.1%

Synonyms: 4-bromo-2-fluoro-1-methoxybenzene

1233000-1 1233000-2 1233000-3
103536-274EA 12.31 USD
103536-274 103536-276 103536-278
4-Bromo-2-fluoroanisole ≥98.1%
4-Bromo-2-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD00011710
CAS Number: 2357-52-0

Order Now

Specification Test Results

Purety 98.1%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR