Pentafluorophenol ≥97.0%

Synonyms: 2,3,4,5,6-Pentafluorophenol

1232946-1 1232946-2 1232946-3
103536-268EA 12.31 USD
103536-268 103536-270 103536-272
Pentafluorophenol ≥97.0%
Pentafluorophenol
Formula: C₆HF₅O
MW: 184.07 g/mol
MDL Number: MFCD00002156
CAS Number: 771-61-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR