3-Fluoro-4-nitroaniline ≥97.0%

1232934-1 1232934-2 1232934-3
103536-262EA 30.77 USD
103536-262 103536-264 103536-266
3-Fluoro-4-nitroaniline ≥97.0%
3-Fluoro-4-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD03412239
CAS Number: 2369-13-3

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR