1-Hydroxy-1-cyclopropanecarboxylic acid ≥95.9%

Synonyms: 1-hydroxycyclopropane-1-carboxylic acid, 1-Hydroxycyclopropanecarboxylic acid

123187-1 123187-2 123187-3
103536-250EA 76.92 USD
103536-250 103536-252 103536-254
1-Hydroxy-1-cyclopropanecarboxylic acid ≥95.9%
1-Hydroxy-1-cyclopropanecarboxylic acid
Formula: C₄H₆O₃
MW: 102.089 g/mol
MDL Number: MFCD00010797
CAS Number: 17994-25-1

Order Now

Specification Test Results

Purety 95.9%
Appearance Off-white crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR