1-Boc-1H-pyrazole-4-boronic acid pinacol ester ≥97.0%

Synonyms: 2-(1-tert-Butoxycarbonyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-Boc-pyrazole-4-boronic acid pinacol ester, tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(tert-Butoxycarbonyl)pyrazolyl-4-boronic acid Pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester, 1-Boc-pyrazolyl-4-boronic acid Pinacol ester, 2-(1-Boc-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-butyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylic acid tert-butyl ester, tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate

122956-1 122956-2 122956-3
103536-244EA 23.08 USD
103536-244 103536-246 103536-248
1-Boc-1H-pyrazole-4-boronic acid pinacol ester ≥97.0%
1-Boc-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₄H₂₃N₂O₄B
MW: 294.1544 g/mol
MDL Number: MFCD05663873
CAS Number: 552846-17-0

Order Now

Specification Test Results

Purety 97.0%
Appearance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR