4-Nitro-1H-pyrazole ≥98.9%

122888-1 122888-2 122888-3
103536-238EA 30.77 USD
103536-238 103536-240 103536-242
4-Nitro-1H-pyrazole ≥98.9%
4-Nitro-1H-pyrazole
Formula: C₃H₃N₃O₂
MW: 113.0800 g/mol
MDL Number: MFCD00159626
CAS Number: 2075-46-9

Order Now

Specification Test Results

Purety 98.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR