Ethyl 5-amino-6-methylpyridine-3-carboxylate ≥97%

Synonyms: Ethyl 5-amino-6-methylnicotinate

118339-1 118339-2 118339-3
103535-316EA 76.92 USD
103535-316 103536-234 103536-236
Ethyl 5-amino-6-methylpyridine-3-carboxylate ≥97%
Ethyl 5-amino-6-methylpyridine-3-carboxylate
Formula: C₉H₁₂N₂O₂
MW: 180.204 g/mol
MDL Number: MFCD18256589
CAS Number: 1008138-73-5

Order Now

Specification Test Results

Purety 97%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR