(1R,4R,5R)-3-Oxo-2-oxabicyclo[2.2.1]heptane-5-carboxylic acid cinchonidine salt ≥97%

117864-1-1 117864-1-2 117864-1-3
103535-310EA 692.31 USD
103535-310 103535-312 103535-314
(1R,4R,5R)-3-Oxo-2-oxabicyclo[2.2.1]heptane-5-carboxylic acid cinchonidine salt ≥97%
(1R,4R,5R)-3-Oxo-2-oxabicyclo[2.2.1]heptane-5-carboxylic acid cinchonidine salt
Formula: C₂₆H₃₀N₂O₅
MW: 450.527 g/mol
CAS Number: 1233219-43-6

Order Now

Specification Test Results

Purety 97%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR