1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride ≥98.2%

Synonyms: [(2,4-dichlorophenyl)methyl](methyl)amine hydrochloride, N-Methyl-2,4-dichlorobenzylamine Hydrochloride

116073-1-1 116073-1-2 116073-1-3
103535-304EA 95.38 USD
103535-304 103535-306 103535-308
1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride ≥98.2%
1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride
Formula: C₈H₁₀Cl₃N
MW: 226.5300 g/mol
MDL Number: MFCD04117880
CAS Number: 90389-07-4

Order Now

Specification Test Results

Purety 98.2%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR