1-Boc-piperidine ≥99.9%

Synonyms: tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine

116071-1 116071-2 116071-3
103535-298EA 30.77 USD
103535-298 103535-300 103535-302
1-Boc-piperidine ≥99.9%
1-Boc-piperidine
Formula: C₁₀H₁₉NO₂
MW: 185.27 g/mol
MDL Number: MFCD02093935
CAS Number: 75844-69-8

Order Now

Specification Test Results

Purety 99.9%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR