5-Bromopyrimidine-2-carbonitrile ≥97.0%

Synonyms: 5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile

113081-1 113081-2 113081-3
103535-292EA 30.77 USD
103535-292 103535-294 103535-296
5-Bromopyrimidine-2-carbonitrile ≥97.0%
5-Bromopyrimidine-2-carbonitrile
Formula: C₅H₂BrN₃
MW: 183.99 g/mol
MDL Number: MFCD02940446
CAS Number: 38275-57-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR