1-(4-Amino-2-fluorophenyl)-4-methylpiperazine ≥97.0%

Synonyms: 3-Fluoro-4-(4-methylpiperazino)aniline, 3-fluoro-4-(4-methyl-1-piperazinyl)benzenamine, 3-Fluoro-4-(4-methylpiperazin-1-yl)aniline

112912-1 112912-2 112912-3
103535-274EA 30.77 USD
103535-274 103535-276 103535-278
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine ≥97.0%
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
Formula: C₁₁H₁₆FN₃
MW: 209.27 g/mol
MDL Number: MFCD05663793
CAS Number: 221198-99-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Pink solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR