4-Chloro-2-iodoaniline ≥97.0%

112897-1 112897-2 112897-3
103535-268EA 12.31 USD
103535-268 103535-270 103535-272
4-Chloro-2-iodoaniline ≥97.0%
4-Chloro-2-iodoaniline
Formula: C₆H₅ClIN
MW: 253.47 g/mol
MDL Number: MFCD01863737
CAS Number: 63069-48-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Purple crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR