2-Aminopyridine ≥97.0%

Synonyms: 2-Pyridylamine, Pyridin-2-amine

112868-1 112868-2 112868-3
103535-262EA 12.31 USD
103535-262 103535-264 103535-266
2-Aminopyridine ≥97.0%
2-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
MDL Number: MFCD00006312
CAS Number: 504-29-0
UN: 2671
ADR: 6.1,II

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR