3-Amino-4-bromo-1H-pyrazole ≥99.2%

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

10010-1 10010-2 10010-3
103535-256EA 38.46 USD
103535-256 103535-258 103535-260
3-Amino-4-bromo-1H-pyrazole ≥99.2%
3-Amino-4-bromo-1H-pyrazole
Formula: C₃H₄BrN₃
MW: 161.99 g/mol
MDL Number: MFCD00082728
CAS Number: 16461-94-2

Order Now

Specification Test Results

Purety 99.2%
Appearance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR