4-Methoxy-1,4'-bipiperidine dihydrochloride ≥99.1%

09431-1-1 09431-1-2 09431-1-3
103535-244EA 323.08 USD
103535-244 103535-246 103535-248
4-Methoxy-1,4'-bipiperidine dihydrochloride ≥99.1%
4-Methoxy-1,4'-bipiperidine dihydrochloride
Formula: C₁₁H₂₂N₂O·2HCl
CAS Number: 930604-26-5

Order Now

Specification Test Results

Purety 99.1%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR