2,6-Dibromopyridine ≥98.4%

08049-1 08049-2 08049-3
103535-232EA 18.46 USD
103535-232 103535-234 103535-236
2,6-Dibromopyridine ≥98.4%
2,6-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
MDL Number: MFCD00006223
CAS Number: 626-05-1

Order Now

Specification Test Results

Purety 98.4%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR