2-Bromo-5-chloropyridine ≥97.8%

08041-1 08041-2 08041-3
103535-226EA 12.31 USD
103535-226 103535-228 103535-230
2-Bromo-5-chloropyridine ≥97.8%
2-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD00234006
CAS Number: 40473-01-6

Order Now

Specification Test Results

Purety 97.8%
Appearance Orange Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR