3,3-Dimethylglutarimide ≥97.0%

Synonyms: 4,4-dimethylpiperidine-2,6-dione

08029-1 08029-2 08029-3
103535-214EA 30.77 USD
103535-214 103535-216 103535-218
3,3-Dimethylglutarimide ≥97.0%
3,3-Dimethylglutarimide
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
MDL Number: MFCD00006671
CAS Number: 1123-40-6

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR