4-Amino-2,6-dichloropyrimidine ≥97.9%

Synonyms: 2,6-Dichloropyrimidin-4-ylamine

08026-1 08026-2 08026-3
103535-208EA 12.31 USD
103535-208 103535-210 103535-212
4-Amino-2,6-dichloropyrimidine ≥97.9%
4-Amino-2,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
MDL Number: MFCD00038015
CAS Number: 10132-07-7

Order Now

Specification Test Results

Purety 97.9%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR