1-(Boc-amino)cyclobutanecarboxylic acid ≥97.0%

07511-1 07511-2 07511-3
103535-202EA 30.77 USD
103535-202 103535-204 103535-206
1-(Boc-amino)cyclobutanecarboxylic acid ≥97.0%
1-(Boc-amino)cyclobutanecarboxylic acid
Formula: C₁₀H₁₇NO₄
MDL Number: MFCD02682623
CAS Number: 120728-10-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR