4-Bromo-2-nitrobenzoic acid ≥97.0%

07456-1 07456-2 07456-3
103535-196EA 12.31 USD
103535-196 103535-198 103535-200
4-Bromo-2-nitrobenzoic acid ≥97.0%
4-Bromo-2-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
MDL Number: MFCD01013599
CAS Number: 99277-71-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR