5-Amino-4,6-dichloro-2-methylpyrimidine ≥99.5%

Synonyms: 4,6-dichloro-2-methylpyrimidin-5-amine

07103-2 07103-3 07103-1
103535-192EA 30.77 USD
103535-192 103535-194 103536-232
5-Amino-4,6-dichloro-2-methylpyrimidine ≥99.5%
5-Amino-4,6-dichloro-2-methylpyrimidine
Formula: C₅H₅Cl₂N₃
MW: 178.0200 g/mol
MDL Number: MFCD00194053
CAS Number: 39906-04-2

Order Now

Specification Test Results

Purety 99.5%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR