Ethyl 6-methyl-5-nitropyridine-3-carboxylate ≥95.2%

Synonyms: Ethyl 6-methyl-5-nitronicotinate

06253-1 06253-2 06253-3
103536-214EA 123.08 USD
103536-214 103536-216 103536-218
Ethyl 6-methyl-5-nitropyridine-3-carboxylate ≥95.2%
Ethyl 6-methyl-5-nitropyridine-3-carboxylate
Formula: C₉H₁₀N₂O₄
MW: 210.187 g/mol
MDL Number: MFCD18260802
CAS Number: 1211538-09-8

Order Now

Specification Test Results

Purety 95.2%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR