(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥98.5%

Synonyms: Boc-D-cis-Hyp-OMe, N-(tert-Butoxycarbonyl)-cis-4-hydroxy-D-proline methyll ester, 1-tert-Butyl 2-Methyl (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-cis-D-Hyp-OMe, 1-tert-butyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate, (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl ester, (2R,4R)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, N-Boc-cis-4-hydroxy-D-proline methyll ester, N-Boc-cis-4-hydroxy-D-proline methyl ester

06230-1 06230-2 06230-3
103536-208EA 38.46 USD
103536-208 103536-210 103536-212
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥98.5%
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
MDL Number: MFCD00797548
CAS Number: 114676-69-6

Order Now

Specification Test Results

Purety 98.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR