3,6-Dihydro-2H-pyran ≥97.8%

06191-1 06191-2 06191-3
103536-196EA 38.46 USD
103536-196 103536-198 103536-200
3,6-Dihydro-2H-pyran ≥97.8%
3,6-Dihydro-2H-pyran
Formula: C₅H₈O
MW: 84.1164 g/mol
MDL Number: MFCD17171194
CAS Number: 3174-74-1

Order Now

Specification Test Results

Purety 97.8%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR