Methyl-4-nitro-1H-pyrazole-3-carboxylate ≥97.7%

Synonyms: Methyl 4-nitro-1H-pyrazole-5-carboxylate, Methyl 4-nitro-1H-pyrazole-3-carboxylate

06094-1 06094-2 06094-3
103536-190EA 23.08 USD
103536-190 103536-192 103536-194
Methyl-4-nitro-1H-pyrazole-3-carboxylate ≥97.7%
Methyl-4-nitro-1H-pyrazole-3-carboxylate
Formula: C₅H₅N₃O₄
MW: 171.11 g/mol
MDL Number: MFCD00505604
CAS Number: 138786-86-4

Order Now

Specification Test Results

Purety 97.7%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR