1H-Pyrazole-4-boronic acid pinacol ester ≥97.0%

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

06068-1 06068-2 06068-3
103536-184EA 107.69 USD
103536-184 103536-186 103536-188
1H-Pyrazole-4-boronic acid pinacol ester ≥97.0%
1H-Pyrazole-4-boronic acid pinacol ester
Formula: C₉H₁₅BN₂O₂
MW: 194.039 g/mol
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR