(3R)-3-Aminobutan-1-ol ≥99.0%

Synonyms: 1-Butanol,3-aMino-, (3R)-, (R)-3-Aminobutanol, (R)-3-Amino-1-butanol, (R)-3-Aminobutan-1-ol, (-)-(R)-3-aMinopropan-1-ol, (3R)-3-Amino-1-butanol, R-3-amino-1-butanol

06001-1 06001-2 06001-3
103536-178EA 19.69 USD
103536-178 103536-180 103536-182
(3R)-3-Aminobutan-1-ol ≥99.0%
(3R)-3-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.1362 g/mol
MDL Number: MFCD13184351
CAS Number: 61477-40-5

Order Now

Specification Test Results

Purety 99.0%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR