(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride ≥97%

Synonyms: (S)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride

05937-01-1 05937-01-2 05937-01-3
103536-172EA 346.15 USD
103536-172 103536-174 103536-176
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride ≥97%
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride
Formula: C₇H₁₄N₂O·2HCl
MW: 215.121 g/mol
MDL Number: MFCD23105798
CAS Number: 1089280-14-7

Order Now

Specification Test Results

Purety 97%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR