1-Boc-piperazine ≥96.9%

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

05908-1 05908-2 05908-3
103535-190EA 12.31 USD
103535-190 103536-168 103536-170
1-Boc-piperazine ≥96.9%
1-Boc-piperazine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD00075265
CAS Number: 57260-71-6
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Purety 96.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR