(S)-Piperazine-2-carboxylic acid dihydrochloride ≥99.2%

Synonyms: (2S)-piperazine-2-carboxylic acid dihydrochloride

05782-01-1 05782-01-2 05782-01-3
103535-172EA 23.08 USD
103535-172 103535-174 103535-176
(S)-Piperazine-2-carboxylic acid dihydrochloride ≥99.2%
(S)-Piperazine-2-carboxylic acid dihydrochloride
Formula: C₅H₁₂Cl₂N₂O₂
MW: 203.07 g/mol
MDL Number: MFCD01632104
CAS Number: 158663-69-5

Order Now

Specification Test Results

Purety 99.2%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR