(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥99.9%

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

05743-1 05743-2 05743-3
103535-166EA 30.77 USD
103535-166 103535-168 103535-170
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥99.9%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
MW: 109.1259 g/mol
MDL Number: MFCD00213364
CAS Number: 49805-30-3

Order Now

Specification Test Results

Purety 99.9%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR