Methyl cis-4-hydroxycyclohexanecarboxylate ≥97.0%

Synonyms: cis-Methyl 4-hydroxycyclohexanecarboxylate

05684-1 05684-2 05684-3
103535-154EA 115.38 USD
103535-154 103535-156 103535-158
Methyl cis-4-hydroxycyclohexanecarboxylate ≥97.0%
Methyl cis-4-hydroxycyclohexanecarboxylate
Formula: C₈H₁₄O₃
MW: 158.197 g/mol
MDL Number: MFCD20441962
CAS Number: 3618-03-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR