5-Bromo-2,4-dichloropyrimidine ≥99.5%

05678-1 05678-2 05678-3
103535-148EA 41.54 USD
103535-148 103535-150 103535-152
5-Bromo-2,4-dichloropyrimidine ≥99.5%
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
MDL Number: MFCD00127818
CAS Number: 36082-50-5

Order Now

Specification Test Results

Purety 99.5%
Appearance Light-Yellow State

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR