trans-4-Aminocyclohexanol ≥97.0%

05677-1 05677-2 05677-3
103535-142EA 12.31 USD
103535-142 103535-144 103535-146
trans-4-Aminocyclohexanol ≥97.0%
trans-4-Aminocyclohexanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00067698
CAS Number: 27489-62-9
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR