Ethyl-4-bromo-1H-pyrazole-3-carboxylate ≥99.2%

Synonyms: Ethyl 4-bromo-1H-pyrazole-3-carboxylate

05672-1 05672-2 05672-3
103535-136EA 76.92 USD
103535-136 103535-138 103535-140
Ethyl-4-bromo-1H-pyrazole-3-carboxylate ≥99.2%
Ethyl-4-bromo-1H-pyrazole-3-carboxylate
Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
MDL Number: MFCD10697501
CAS Number: 5932-34-3

Order Now

Specification Test Results

Purety 99.2%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR