Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate ≥99.3%

Synonyms: Benzyl 5,6-dihydropyridine-1(2H)-carboxylate

05654-1 05654-2 05654-3
103535-130EA 30.77 USD
103535-130 103535-132 103535-134
Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate ≥99.3%
Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate
Formula: C₁₃H₁₅NO₂
MW: 217.264 g/mol
MDL Number: MFCD07366998
CAS Number: 66207-23-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Purety 99.3%
Appearance Colorless oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR