1,2-O-Isopropylidene-ɑ-D-xylofuranose ≥97.0%

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

05522-1 05522-2 05522-3
103535-112EA 23.08 USD
103535-112 103535-114 103535-116
1,2-O-Isopropylidene-ɑ-D-xylofuranose ≥97.0%
1,2-O-Isopropylidene-alpha-D-xylofuranose
Formula: C₈H₁₄O₅
MW: 190.20 g/mol
MDL Number: MFCD00063295
CAS Number: 20031-21-4

Order Now

Specification Test Results

Purety 97.0%
Appearance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR