Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate ≥98.2%

Synonyms: ethyl 2-(trifluoromethyl)-1,3-thiazole-5-carboxylate, Ethyl 2-(trifluoromethyl)thiazole-5-carboxylate

05500-1 05500-2 05500-3
103535-106EA 61.54 USD
103535-106 103535-108 103535-110
Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate ≥98.2%
Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate
Formula: C₇H₆F₃NO₂S
MW: 225.1883 g/mol
MDL Number: MFCD09055619
CAS Number: 131748-96-4

Order Now

Specification Test Results

Purety 98.2%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR