tert-Butyl 4-(bromomethyl)-4-fluoropiperidine-1-carboxylate ≥97%

05419-1 05419-2 05419-3
103536-162EA 76.92 USD
103536-162 103536-164 103536-166
tert-Butyl 4-(bromomethyl)-4-fluoropiperidine-1-carboxylate ≥97%
tert-Butyl 4-(bromomethyl)-4-fluoropiperidine-1-carboxylate
Formula: C₁₁H₁₉BrFNO₂
MW: 296.176 g/mol
MDL Number: MFCD11973737
CAS Number: 1207176-24-6

Order Now

Specification Test Results

Purety 97%
Appearance Off-white crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR