3-Chloro-5-(trifluoromethyl)pyridine ≥98.0%

05379-1 05379-2 05379-3
103536-156EA 36.92 USD
103536-156 103536-158 103536-160
3-Chloro-5-(trifluoromethyl)pyridine ≥98.0%
3-Chloro-5-(trifluoromethyl)pyridine
Formula: C6H3ClF3N
MW: 181.55 g/mol
MDL Number: MFCD00042227
CAS Number: 85148-26-1

Order Now

Specification Test Results

Purety 98.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR