2,6-Dichloro-3-cyano-4-picoline ≥99.3%

Synonyms: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine

05226-1 05226-2 05226-3
103536-150EA 12.31 USD
103536-150 103536-152 103536-154
2,6-Dichloro-3-cyano-4-picoline ≥99.3%
2,6-Dichloro-3-cyano-4-picoline
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00052633
CAS Number: 875-35-4

Order Now

Specification Test Results

Purety 99.3%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR