Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate ≥99.0%

Synonyms: (S)-Isopropyl 2-(((S)-(perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate

05156-1 05156-2 05156-3
103536-144EA 12.31 USD
103536-144 103536-146 103536-148
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate ≥99.0%
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate
Formula: C₁₈H₁₇F₅NO₅
MW: 453.298 g/mol
MDL Number: MFCD22665799
CAS Number: 1334513-02-8

Order Now

Specification Test Results

Purety 99.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR