4,5-Dichloro-3(2H)-pyridazinone ≥90.7%

Synonyms: 4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one

05079-1 05079-2 05079-3
103536-138EA 18.46 USD
103536-138 103536-140 103536-142
4,5-Dichloro-3(2H)-pyridazinone ≥90.7%
4,5-Dichloro-3(2H)-pyridazinone
Formula: C₄H₂Cl₂N₂O
MW: 164.98 g/mol
MDL Number: MFCD00051504
CAS Number: 932-22-9

Order Now

Specification Test Results

Purety 90.7%
Appearance White floc

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR