tert-Butyl 3-hydroxy-3-methylazetidine-1-carboxylate ≥99.3%

05003-1 05003-2 05003-3
103535-094EA 30.77 USD
103535-094 103535-096 103535-098
tert-Butyl 3-hydroxy-3-methylazetidine-1-carboxylate ≥99.3%
tert-Butyl 3-hydroxy-3-methylazetidine-1-carboxylate
Formula: C₉H₁₇NO₃
MW: 187.236 g/mol
MDL Number: MFCD16037112
CAS Number: 1104083-23-9

Order Now

Specification Test Results

Purety 99.3%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR