2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide ≥97.0%

Synonyms: 2,2,2-trifluoro-N-{6-iodoimidazo[1,2-a]pyridin-2-yl}acetamide

03900-1 03900-2 03900-3
103535-082EA 243.08 USD
103535-082 103535-084 103535-086
2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide ≥97.0%
2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide
Formula: C₉H₅F₃IN₃O
MW: 355.06 g/mol
MDL Number: MFCD11878598
CAS Number: 209971-49-3

Order Now

Specification Test Results

Purety 97.0%
Appearance Pink solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR