tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride ≥98.7%

Synonyms: tert-Butyl azetidin-3-yl(methyl)carbamate hydrochloride

03881-01-1 03881-01-2 03881-01-3
103535-076EA 38.46 USD
103535-076 103535-078 103535-080
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride ≥98.7%
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride
Formula: C₉H₁₉ClN₂O₂
MW: 222.712 g/mol
MDL Number: MFCD12963843
CAS Number: 943060-59-1

Order Now

Specification Test Results

Purety 98.7%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR