(±)-Cbz-α-phosphonoglycine trimethyl ester ≥97.0%

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

03879-1 03879-2 03879-3
103536-132EA 18.46 USD
103536-132 103536-134 103536-136
(±)-Cbz-α-phosphonoglycine trimethyl ester ≥97.0%
(±)-Cbz-α-phosphonoglycine trimethyl ester
Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
MDL Number: MFCD00043304
CAS Number: 88568-95-0

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR